Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003128.2 | NP_003119.2 | 15 | 3016 | ||
NM_178313.2 | NP_842565.2 | 14 | 3113 | |||
XM_005264517.2 | XP_005264574.1 | 15 | 2925 | |||
XM_005264518.1 | XP_005264575.1 | 14 | 3771 | |||
XM_006712087.2 | XP_006712150.1 | 15 | 2889 | |||
XM_017004779.1 | XP_016860268.1 | 15 | 3032 | |||
XM_017004780.1 | XP_016860269.1 | 15 | 2986 | |||
XM_017004781.1 | XP_016860270.1 | 15 | 2987 | |||
GenBank mRNA | AB209748.1 | - | 15 | 2894 | ||
AB371586.1 | - | 15 | 3006 | |||
AF327441.1 | - | 14 | 3113 | |||
BC137282.1 | - | 14 | 3023 | 9021279 | ||
BC137283.1 | - | 14 | 3023 | 9021280 | ||
EU794662.1 | - | 15 | 3016 | |||
JQ599728.1 | - | 14 | 2767 | |||
S65762.1 | - | 7 | 1891 |