Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000348.3 | NP_000339.2 | 5 | 818 | ||
XM_011533069.2 | XP_011531371.1 | 5 | 1533 | |||
XM_011533072.2 | XP_011531374.1 | 8 | 963 | |||
GenBank mRNA | AK307235.1 | - | 5 | 554 | ||
AK316597.1 | - | 5 | 788 | |||
BC112252.1 | - | 5 | 805 | 8327720 | ||
BC113641.1 | - | 5 | 793 | 8322693 | ||
EF560740.1 | - | 5 | 786 | |||
M74047.1 | - | 5 | 774 |