Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003141.3 | NP_003132.2 | 5, 6 | 877 | ||
GenBank mRNA | AY742713.1 | - | 4, 5 | 764 | ||
BC010861.1 | - | 5, 6 | 848 | 3900540 | ||
M34551.1 | - | 4, 5 | 802 | |||
M62800.1 | - | 5, 6 | 849 |