Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256510.1 | NP_001243439.1 | 4, 5 | 650 | ||
NM_001256511.1 | NP_001243440.1 | 4, 5 | 531 | |||
NM_001256512.1 | NP_001243441.1 | 4, 5 | 513 | |||
NM_001256513.1 | NP_001243442.1 | 4, 5 | 441 | |||
NM_003143.2 | NP_003134.1 | 4, 5 | 348 | |||
NR_046269.1 | 5, 6 | 398 | ||||
XM_005250048.4 | XP_005250105.1 | 4, 5 | 360 | |||
XM_005250049.4 | XP_005250106.1 | 4, 5 | 399 | |||
XM_005250050.4 | XP_005250107.1 | 4, 5 | 450 | |||
XM_005250051.4 | XP_005250108.1 | 4, 5 | 476 | |||
GenBank mRNA | AF277319.1 | - | 4, 5 | 294 | ||
AK294393.1 | - | 4, 5 | 304 | |||
AK313033.1 | - | 4, 5 | 361 | |||
BC000895.1 | - | 4, 5 | 650 | 3464282 | ||
BC093054.1 | - | 4, 5 | 310 | 30722138 | ||
BF131408.1 | - | 5, 6 | 328 | |||
BG030060.1 | - | 464 | ||||
BP308841.1 | - | 3, 4 | 217 | |||
BQ959702.1 | - | 4, 5 | 413 | |||
BU532904.1 | - | 4, 5 | 351 | |||
M94556.1 | - | 4, 5 | 293 |