Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048.3 | NP_001039.1 | 2 | 302 | ||
GenBank mRNA | AK312177.1 | - | 2 | 281 | ||
BC032625.1 | - | 2 | 207 | 5240781 | ||
BM353331.1 | - | 1 | 363 |