Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005419.3 | NP_005410.1 | 14 | 1444 | ||
NM_198332.1 | NP_938146.1 | 14 | 1432 | |||
XM_005269111.3 | XP_005269168.2 | 14 | 1352 | |||
XM_011538697.2 | XP_011536999.1 | 14 | 1352 | |||
XM_011538698.2 | XP_011537000.1 | 14 | 1313 | |||
XM_011538699.2 | XP_011537001.1 | 14 | 1352 | |||
XM_011538700.2 | XP_011537002.1 | 8 | 750 | |||
XM_017019904.1 | XP_016875393.1 | 13 | 1811 | |||
GenBank mRNA | AK296939.1 | - | 14 | 1305 | ||
AK296940.1 | - | 14 | 1305 | |||
AK308051.1 | - | 8 | 728 | |||
BC006092.1 | - | 13 | 1231 | |||
BC051284.1 | - | 13 | 1264 | 6460486 | ||
BX640607.1 | - | 1 | 171 | |||
M97934.1 | - | 14 | 1278 |