Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 16 | 1655 | ||
NM_003151.3 | NP_003142.1 | 16 | 1706 | |||
XM_006712719.3 | XP_006712782.1 | 16 | 1701 | |||
XM_011511705.2 | XP_011510007.1 | 19 | 1852 | |||
XM_017004784.1 | XP_016860273.1 | 16 | 1460 | |||
GenBank mRNA | AB209043.1 | - | 6 | 342 | ||
AK299976.1 | - | 13 | 1187 | |||
AK300868.1 | - | 16 | 1459 | |||
AK301172.1 | - | 14 | 1130 | |||
BC031212.1 | - | 16 | 1627 | 4830583 | ||
L78440.1 | - | 16 | 1472 |