Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STAU1 SIRNA Assays ›
Gene Symbol:
STAU1
Gene Name:
staufen double-stranded RNA binding protein 1
Gene Aliases:
PPP1R150, STAU
Chromosome Location:
Chr.20: 49113339 - 49219295 on Build GRCh38
UniGene ID:
Hs.596704
Species:
Human
siRNA ID
HSS110295
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STAU1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037328.2
NP_001032405.1
5
657
NM_001319134.1
NP_001306063.1
4
534
NM_001319135.1
NP_001306064.1
6
734
NM_001322927.1
NP_001309856.1
3
459
NM_001322928.1
NP_001309857.1
6
803
NM_001322929.1
NP_001309858.1
6
946
NM_001322930.1
NP_001309859.1
4
748
NM_001322931.1
NP_001309860.1
7
926
NM_001322932.1
NP_001309861.1
4
748
NM_001322933.1
NP_001309862.1
5
823
NM_004602.3
NP_004593.2
5
657
NM_017452.3
NP_059346.2
4
534
NM_017453.3
NP_059347.2
5
823
NM_017454.3
NP_059348.2
3
459
XM_005260527.1
XP_005260584.1
5
706
XM_006723865.1
XP_006723928.1
6
943
XM_011529015.1
XP_011527317.1
7
1056
XM_017028028.1
XP_016883517.1
6
800
GenBank mRNA
AB209561.1
-
5
688
AF061938.1
-
4
528
AF061939.1
-
5
817
AF061940.1
-
3
453
AF061941.1
-
5
660
AJ132258.1
-
2
201
AK025519.1
-
4
497
AK057739.1
-
5
780
AK291487.1
-
5
738
AK292859.1
-
4
319
AL136601.2
-
4
517
AY529074.1
-
4
352
AY546099.1
-
4
528
BC001893.1
-
5
461
BC010169.2
-
5
461
BC050432.1
-
4
561
6047265
BC095397.1
-
4
506
30528051
DB086071.1
-
5
446
DB100321.1
-
4
533
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to oxidative stress
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
positive regulation of long-term synaptic potentiation
double-stranded RNA binding
protein binding
protein phosphatase 1 binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline