Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005990.3 | NP_005981.3 | 13, 14 | 2383 | ||
XM_017009788.1 | XP_016865277.1 | 12, 13 | 1937 | |||
GenBank mRNA | AB015718.1 | - | 13, 14 | 2133 | ||
AK022960.1 | - | 5, 6 | 956 | |||
AK307943.1 | - | 13, 14 | 1917 | |||
AK313350.1 | - | 14, 15 | 2138 | |||
BC070077.1 | - | 13, 14 | 2297 | 5267449 |