Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032221.3 | NP_001027392.1 | 5, 6 | 529 | ||
NM_003165.3 | NP_003156.1 | 5, 6 | 529 | |||
GenBank mRNA | AF004562.1 | - | 5, 6 | 452 | ||
AF004563.1 | - | 5, 6 | 452 | |||
AK293984.1 | - | 4, 5 | 348 | |||
AK295154.1 | - | 4, 5 | 387 | |||
BC015749.1 | - | 5, 6 | 445 | 4868854 | ||
D63851.1 | - | 5, 6 | 401 | |||
Y12723.1 | - | 5, 6 | 440 |