Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001056.3 | NP_001047.1 | 6 | 1005 | ||
NM_176825.2 | NP_789795.1 | 7 | 1038 | |||
XM_005264013.3 | XP_005264070.1 | 6 | 826 | |||
GenBank mRNA | AB008164.1 | - | 6 | 675 | ||
AF026303.1 | - | 6 | 637 | |||
AF186251.1 | - | 6 | 870 | |||
AF186252.1 | - | 6 | 1005 | |||
AF186253.1 | - | 6 | 721 | |||
AF186254.1 | - | 7 | 903 | |||
AF186255.1 | - | 7 | 1038 | |||
AF186256.1 | - | 7 | 754 | |||
AI986284.1 | - | 3 | 612 | |||
AK297086.1 | - | 6 | 811 | |||
AK298306.1 | - | 6 | 676 | |||
AK313193.1 | - | 6 | 769 | |||
BC005353.1 | - | 6 | 622 | 4051264 | ||
BT006951.1 | - | 5 | 552 | |||
U66036.1 | - | 6 | 728 |