Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003167.3 | NP_003158.2 | 2 | 332 | ||
GenBank mRNA | AK289380.1 | - | 2 | 274 | ||
AK313415.1 | - | 2 | 259 | |||
BC020755.1 | - | 2 | 249 | 4733628 | ||
BG563083.1 | - | 2 | 312 | |||
BX509490.1 | - | 2 | 333 | |||
L02337.1 | - | 2 | 257 | |||
L20000.1 | - | 2 | 244 | |||
S43859.1 | - | 2 | 257 | |||
S43861.1 | - | 1 | 26 | |||
U08024.1 | - | 2 | 212 | |||
U08025.1 | - | 2 | 256 | |||
X70222.1 | - | 2 | 244 | |||
X84816.1 | - | 2 | 237 |