Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003167.3 | NP_003158.2 | 5 | 772 | ||
GenBank mRNA | AK289380.1 | - | 5 | 714 | ||
AK313415.1 | - | 5 | 699 | |||
BC020755.1 | - | 5 | 689 | 4733628 | ||
L02337.1 | - | 5 | 697 | |||
L20000.1 | - | 5 | 684 | |||
S43859.1 | - | 5 | 697 | |||
S43861.1 | - | 4 | 466 | |||
U08024.1 | - | 5 | 652 | |||
U08025.1 | - | 5 | 696 | |||
X70222.1 | - | 5 | 684 | |||
X84816.1 | - | 5 | 677 |