Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280788.1 | NP_001267717.1 | 2 | 262 | ||
NM_001280789.1 | NP_001267718.1 | 2 | 407 | |||
NM_001280790.1 | NP_001267719.1 | 3 | 557 | |||
NM_001280791.1 | NP_001267720.1 | 2 | 505 | |||
NM_001280792.1 | NP_001267721.1 | 2 | 407 | |||
NM_001280793.1 | NP_001267722.1 | 2 | 407 | |||
NM_033161.3 | NP_149351.1 | 2 | 407 | |||
XM_011518943.1 | XP_011517245.1 | 4 | 819 | |||
XM_011518944.2 | XP_011517246.1 | 5 | 753 | |||
XM_011518945.2 | XP_011517247.1 | 2 | 2582 | |||
XM_017015062.1 | XP_016870551.1 | 5 | 829 | |||
GenBank mRNA | AF078866.1 | - | 2 | 323 | ||
AF078867.1 | - | 2 | 323 | |||
AK293434.1 | - | 2 | 339 | |||
AK295435.1 | - | 3 | 454 | |||
AK299656.1 | - | 2 | 301 | |||
AK300004.1 | - | 2 | 301 | |||
AK301737.1 | - | 5 | 753 | |||
AK303309.1 | - | 2 | 271 | |||
AK310333.1 | - | 5 | 829 | |||
AK315488.1 | - | 2 | 361 | |||
AK316480.1 | - | 2 | 399 | |||
BC018741.1 | - | 2 | 230 | 5014698 | ||
BC111023.1 | - | 2 | 272 | 5203634 | ||
BM148022.1 | - | 2 | 341 | |||
BM557312.1 | - | 2 | 261 | |||
BX647961.1 | - | 2 | 260 | |||
DA842525.1 | - | 2 | 294 | |||
DC424374.1 | - | 2 | 409 | |||
Y14820.1 | - | 2 | 189 |