Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280788.1 | NP_001267717.1 | 6 | 848 | ||
NM_001280790.1 | NP_001267719.1 | 7 | 1143 | |||
NM_001280791.1 | NP_001267720.1 | 6 | 1091 | |||
NM_001280792.1 | NP_001267721.1 | 5 | 806 | |||
NM_001280793.1 | NP_001267722.1 | 5 | 916 | |||
NM_033161.3 | NP_149351.1 | 6 | 993 | |||
XM_011518943.1 | XP_011517245.1 | 8 | 1405 | |||
XM_011518944.2 | XP_011517246.1 | 9 | 1339 | |||
XM_011518945.2 | XP_011517247.1 | 5 | 2981 | |||
XM_017015062.1 | XP_016870551.1 | 8 | 1228 | |||
GenBank mRNA | AF078866.1 | - | 6 | 909 | ||
AK293434.1 | - | 6 | 925 | |||
AK295435.1 | - | 7 | 1040 | |||
AK299656.1 | - | 5 | 722 | |||
AK300004.1 | - | 5 | 700 | |||
AK301737.1 | - | 9 | 1339 | |||
AK303309.1 | - | 6 | 857 | |||
AK315488.1 | - | 6 | 947 | |||
AK316480.1 | - | 6 | 985 | |||
BC018741.1 | - | 6 | 816 | 5014698 | ||
BC111023.1 | - | 6 | 858 | 5203634 | ||
BX647961.1 | - | 6 | 846 |