Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003179.2 | NP_003170.1 | 5 | 534 | ||
GenBank mRNA | AK295524.1 | - | 4 | 400 | ||
AK313030.1 | - | 5 | 519 | |||
AK315953.1 | - | 4 | 400 | |||
BC064550.1 | - | 5 | 534 | 5768012 | ||
X06389.1 | - | 4 | 467 |