Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297774.1 | NP_001284703.1 | 5 | 1037 | ||
NM_003180.2 | NP_003171.2 | 7 | 1092 | |||
XM_006723339.3 | XP_006723402.1 | 7 | 927 | |||
XM_006723340.3 | XP_006723403.1 | 7 | 1178 | |||
XM_006723341.3 | XP_006723404.1 | 5 | 1511 | |||
XM_017027175.1 | XP_016882664.1 | 7 | 989 | |||
XM_017027176.1 | XP_016882665.1 | 7 | 980 | |||
GenBank mRNA | AK125182.1 | - | 6 | 1880 | ||
AK295337.1 | - | 5 | 984 | |||
AK313067.1 | - | 7 | 881 | |||
BC029962.1 | - | 7 | 1020 | |||
BC033683.1 | - | 7 | 896 | |||
BC046157.1 | - | 7 | 1018 | 5768093 | ||
DQ090951.1 | - | 6 | 808 |