Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297774.1 | NP_001284703.1 | 3 | 613 | ||
NM_003180.2 | NP_003171.2 | 5 | 665 | |||
XM_006723339.3 | XP_006723402.1 | 5 | 500 | |||
XM_006723340.3 | XP_006723403.1 | 5 | 754 | |||
XM_006723341.3 | XP_006723404.1 | 3 | 1084 | |||
XM_017027175.1 | XP_016882664.1 | 5 | 562 | |||
XM_017027176.1 | XP_016882665.1 | 5 | 553 | |||
GenBank mRNA | AK125182.1 | - | 4 | 1453 | ||
AK295337.1 | - | 3 | 560 | |||
AK313067.1 | - | 5 | 454 | |||
BC029962.1 | - | 5 | 593 | |||
BC033683.1 | - | 5 | 472 | |||
BC046157.1 | - | 5 | 591 | 5768093 | ||
BQ417474.1 | - | 5 | 578 | |||
DQ090951.1 | - | 4 | 384 | |||
Y19236.1 | - | 3 | 183 |