Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003185.3 | NP_003176.2 | 15 | 3180 | ||
GenBank mRNA | AK097744.1 | - | 3 | 1760 | ||
AK123937.1 | - | 11 | 2029 | |||
U75308.1 | - | 17 | 2766 | |||
Y11354.1 | - | 15 | 3174 |