Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000593.5 | NP_000584.2 | 2 | 1001 | ||
NM_001292022.1 | NP_001278951.1 | 2 | 261 | |||
GenBank mRNA | AB012644.1 | - | 2 | 665 | ||
AK302340.1 | - | 2 | 232 | |||
AK313762.1 | - | 2 | 1037 | |||
AY523970.1 | - | 2 | 872 | |||
AY523971.2 | - | 2 | 872 | |||
BC014081.2 | - | 2 | 1001 | 4304716 | ||
BX648013.1 | - | 2 | 1009 | |||
DC354917.1 | - | 2 | 261 | |||
KU178346.1 | - | 2 | 846 | |||
L21204.1 | - | 2 | 666 | |||
L21205.1 | - | 2 | 666 | |||
L21206.1 | - | 2 | 666 | |||
L21207.1 | - | 2 | 666 | |||
L21208.1 | - | 2 | 666 | |||
X57521.1 | - | 2 | 300 | |||
X57522.1 | - | 2 | 876 |