Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005646.3 | NP_005637.3 | 24 | 3870 | ||
XM_005273234.3 | XP_005273291.1 | 24 | 3867 | |||
XM_011544263.2 | XP_011542565.1 | 23 | 3865 | |||
XM_017002195.1 | XP_016857684.1 | 21 | 2743 | |||
XM_017002196.1 | XP_016857685.1 | 12 | 1715 | |||
XM_017002197.1 | XP_016857686.1 | 19 | 2483 | |||
GenBank mRNA | AK055044.1 | - | 1 | 1938 | ||
BC016023.1 | - | 1 | 363 | |||
U38847.1 | - | 24 | 3870 |