Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005646.3 | NP_005637.3 | 27 | 4417 | ||
XM_005273234.3 | XP_005273291.1 | 27 | 4414 | |||
XM_011544263.2 | XP_011542565.1 | 26 | 4412 | |||
XM_017002195.1 | XP_016857684.1 | 24 | 3290 | |||
XM_017002196.1 | XP_016857685.1 | 15 | 2262 | |||
XM_017002197.1 | XP_016857686.1 | 22 | 3030 | |||
GenBank mRNA | AK055044.1 | - | 4 | 2409 | ||
BC016023.1 | - | 5 | 1006 | |||
U38847.1 | - | 27 | 4417 |