Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079515.2 | NP_001072983.1 | 6 | 765 | ||
NM_001287801.1 | NP_001274730.1 | 6 | 717 | |||
NM_001287802.1 | NP_001274731.1 | 5 | 688 | |||
NM_003193.4 | NP_003184.1 | 6 | 717 | |||
GenBank mRNA | AK093397.1 | - | 5 | 593 | ||
AK292287.1 | - | 6 | 622 | |||
AK296185.1 | - | 6 | 647 | |||
AK298050.1 | - | 6 | 703 | |||
BC008654.1 | - | 6 | 609 | 3872809 | ||
BT007086.1 | - | 5 | 541 | |||
U61232.1 | - | 6 | 670 |