Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000354.5 | NP_000345.2 | 4 | 1077 | ||
XM_005262180.3 | XP_005262237.1 | 4 | 1075 | |||
XM_006724683.2 | XP_006724746.1 | 4 | 1075 | |||
GenBank mRNA | BC020747.2 | - | 3 | 1046 | 4716693 | |
M14091.1 | - | 3 | 1360 |