Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172085.1 | NP_001165556.1 | 5 | 865 | ||
NM_003194.4 | NP_003185.1 | 6 | 1067 | |||
GenBank mRNA | AK298272.1 | - | 7 | 949 | ||
AK304648.1 | - | 5 | 852 | |||
AK315222.1 | - | 6 | 1030 | |||
BC109053.1 | - | 6 | 978 | 40032492 | ||
BC109054.1 | - | 7 | 832 | 40032493 | ||
BC110341.1 | - | 6 | 1026 | 5761426 | ||
BT019657.1 | - | 5 | 779 | |||
CR456776.1 | - | 5 | 788 | |||
M34960.1 | - | 5 | 788 | |||
M55654.1 | - | 6 | 1029 | |||
X54993.1 | - | 5 | 874 |