Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBXAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001061.4 | NP_001052.2 | 4 | 487 | ||
NM_001130966.2 | NP_001124438.1 | 8 | 790 | |||
NM_001166253.1 | NP_001159725.1 | 4 | 487 | |||
NM_001166254.1 | NP_001159726.1 | 6 | 537 | |||
NM_001314028.1 | NP_001300957.1 | 4 | 516 | |||
NM_030984.3 | NP_112246.2 | 4 | 487 | |||
XM_011516544.2 | XP_011514846.1 | 4 | 422 | |||
XM_017012570.1 | XP_016868059.1 | 4 | 422 | |||
XM_017012571.1 | XP_016868060.1 | 4 | 422 | |||
XM_017012572.1 | XP_016868061.1 | 4 | 422 | |||
GenBank mRNA | AK223466.1 | - | 8 | 638 | ||
AK296069.1 | - | 4 | 387 | |||
AK301049.1 | - | 4 | 329 | |||
AK301164.1 | - | 4 | 400 | |||
AK303440.1 | - | 2 | 219 | |||
BC014117.2 | - | 6 | 537 | 4548935 | ||
BC041157.1 | - | 4 | 403 | 5755195 | ||
M74055.1 | - | 4 | 204 | |||
M80646.1 | - | 4 | 423 | |||
M80647.1 | - | 4 | 423 | |||
S60133.1 | - | 4 | 204 |