Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 6 | 1411 | ||
NM_001306179.1 | NP_001293108.1 | 6 | 1411 | |||
XM_005253931.3 | XP_005253988.1 | 6 | 1464 | |||
GenBank mRNA | AK298422.1 | - | 6 | 1285 | ||
BC104908.1 | - | 6 | 1230 | 8143911 | ||
BC104910.1 | - | 6 | 1230 | 8143913 | ||
BC143483.1 | - | 6 | 1231 | |||
HM116541.1 | - | 6 | 1353 | |||
HM116542.1 | - | 6 | 1027 | |||
HM116543.1 | - | 4 | 816 | |||
HM116545.1 | - | 3 | 340 | |||
HM116547.1 | - | 6 | 1353 | |||
HM116552.1 | - | 7 | 970 | |||
HM116557.1 | - | 5 | 1141 | |||
HM116558.1 | - | 5 | 1086 | |||
HM116559.1 | - | 2 | 281 | |||
HM116560.1 | - | 3 | 205 | |||
HM116563.1 | - | 2 | 217 | |||
HM449088.1 | - | 6 | 1233 | |||
HM449089.1 | - | 5 | 1033 | |||
M57732.1 | - | 6 | 1233 | |||
X71346.1 | - | 6 | 1406 | |||
X71347.1 | - | 6 | 1341 |