Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001062.3 | NP_001053.2 | 5, 6 | 855 | ||
GenBank mRNA | AK292990.1 | - | 5, 6 | 772 | ||
BC018632.1 | - | 5, 6 | 777 | |||
BM769693.1 | - | 1, 2 | 101 | |||
J05068.1 | - | 5, 6 | 832 |