Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 5 | 1173 | ||
NM_001290077.1 | NP_001277006.1 | 5 | 1173 | |||
NM_001290078.1 | NP_001277007.1 | 4 | 861 | |||
XM_005251561.1 | XP_005251618.1 | 5 | 840 | |||
XM_005251563.1 | XP_005251620.1 | 5 | 840 | |||
GenBank mRNA | AB086825.1 | - | 5 | 829 | ||
AB208796.1 | - | 5 | 851 | |||
AK291775.1 | - | 5 | 861 | |||
AK294887.1 | - | 4 | 862 | |||
AK295043.1 | - | 5 | 899 | |||
AK308447.1 | - | 3 | 420 | |||
BC035514.1 | - | 5 | 848 | 5228999 | ||
EU826591.1 | - | 5 | 723 | |||
EU826592.1 | - | 5 | 723 | |||
L06139.1 | - | 5 | 871 |