Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003223.2 | NP_003214.1 | 1 | 297 | ||
GenBank mRNA | AK309301.1 | - | 1 | 364 | ||
BC010576.1 | - | 1 | 186 | 4181538 | ||
BU159415.1 | - | 1 | 250 | |||
CR541726.1 | - | 1 | 43 | |||
DA449763.1 | - | 1 | 364 | |||
S73885.1 | - | 1 | 301 |