Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005654.5 | NP_005645.1 | 3 | 2926 | ||
XM_017009797.1 | XP_016865286.1 | 3 | 919 | |||
GenBank mRNA | BC004154.2 | - | 3 | 1327 | 2824138 | |
BC017493.1 | - | 3 | 2926 | 3912075 | ||
HQ709180.1 | - | 4 | 1161 | |||
HQ709181.1 | - | 4 | 1086 | |||
HQ709182.1 | - | 4 | 1116 | |||
HQ709183.1 | - | 4 | 1164 | |||
X16155.1 | - | 3 | 1224 |