Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005423.4 | NP_005414.1 | 3 | 406 | ||
GenBank mRNA | AA307808.1 | - | 3 | 406 | ||
BC032820.1 | - | 3 | 285 | 5184770 | ||
BM817851.1 | - | 3 | 305 | |||
BM853946.1 | - | 3 | 303 |