Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206855.1 | NP_001193784.1 | 9 | 1118 | ||
NM_003227.3 | NP_003218.2 | 12 | 1538 | |||
XM_005250553.4 | XP_005250610.1 | 13 | 1844 | |||
XM_017012573.1 | XP_016868062.1 | 13 | 1646 | |||
GenBank mRNA | AF067864.1 | - | 12 | 1524 | ||
AK000421.1 | - | 8 | 850 | |||
BC142630.1 | - | 12 | 1524 | 40147690 | ||
BC150192.1 | - | 9 | 1332 | |||
BI917390.1 | - | 5 | 570 |