Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128148.2 | NP_001121620.1 | 16 | 1797 | ||
NM_001313965.1 | NP_001300894.1 | 15 | 1595 | |||
NM_001313966.1 | NP_001300895.1 | 14 | 1399 | |||
NM_003234.3 | NP_003225.2 | 16 | 1938 | |||
GenBank mRNA | AB209254.1 | - | 16 | 1701 | ||
AK291723.1 | - | 16 | 1938 | |||
AK294758.1 | - | 14 | 1329 | |||
BC001188.1 | - | 16 | 1711 | 3354176 | ||
BX537966.1 | - | 15 | 1525 | |||
M11507.1 | - | 16 | 1750 | |||
X01060.1 | - | 16 | 1918 |