Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024847.2 | NP_001020018.1 | 5 | 1049 | ||
NM_003242.5 | NP_003233.4 | 4 | 974 | |||
XM_011534043.2 | XP_011532345.1 | 5 | 822 | |||
XM_011534045.2 | XP_011532347.1 | 4 | 602 | |||
XM_017007106.1 | XP_016862595.1 | 4 | 2188 | |||
GenBank mRNA | AJ786388.1 | - | 3 | 288 | ||
AK300383.1 | - | 5 | 950 | |||
AK304404.1 | - | 5 | 719 | |||
AK314102.1 | - | 4 | 890 | |||
BC040499.1 | - | 4 | 975 | |||
D28131.1 | - | 5 | 607 | |||
KC792576.1 | - | 4 | 631 | |||
KC792577.1 | - | 4 | 631 | |||
KC792578.1 | - | 4 | 631 | |||
KC792579.1 | - | 4 | 631 | |||
KC792580.1 | - | 4 | 631 | |||
KU178359.1 | - | 4 | 592 | |||
KU178360.1 | - | 5 | 667 | |||
M85079.1 | - | 4 | 927 |