Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000359.2 | NP_000350.1 | 4 | 727 | ||
GenBank mRNA | AK291350.1 | - | 4 | 694 | ||
AK299436.1 | - | 4 | 496 | |||
AK315819.1 | - | 4 | 727 | |||
AK315828.1 | - | 4 | 687 | |||
AK315843.1 | - | 4 | 694 | |||
BC034699.1 | - | 4 | 678 | 4747045 | ||
D90287.1 | - | 4 | 683 | |||
M55183.1 | - | 4 | 656 | |||
M62925.1 | - | 4 | 689 |