Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003246.3 | NP_003237.2 | 15 | 2536 | ||
XM_011521971.1 | XP_011520273.1 | 14 | 2358 | |||
GenBank mRNA | AB209912.1 | - | 15 | 2522 | ||
AK291639.1 | - | 15 | 2536 | |||
AK304754.1 | - | 15 | 2282 | |||
AK310820.1 | - | 5 | 1071 | |||
BC136469.1 | - | 15 | 2510 | 9020458 | ||
BC136470.1 | - | 15 | 2510 | 9020459 | ||
X04665.1 | - | 15 | 2432 | |||
X14787.1 | - | 15 | 2468 |