Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003246.3 | NP_003237.2 | 3 | 461 | ||
XM_011521971.1 | XP_011520273.1 | 3 | 457 | |||
GenBank mRNA | AB209912.1 | - | 3 | 447 | ||
AK291639.1 | - | 3 | 461 | |||
AU279999.1 | - | 3 | 463 | |||
BC028145.1 | - | 2 | 316 | |||
BC136469.1 | - | 3 | 435 | 9020458 | ||
BC136470.1 | - | 3 | 435 | 9020459 | ||
M14326.1 | - | 3 | 402 | |||
M25631.1 | - | 3 | 381 | |||
X04665.1 | - | 3 | 357 | |||
X14787.1 | - | 3 | 393 |