Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252607.1 | NP_001239536.1 | 13 | 1669 | ||
NM_001252608.1 | NP_001239537.1 | 12 | 1336 | |||
NM_007112.4 | NP_009043.1 | 14 | 1696 | |||
NR_045553.1 | 13 | 1680 | ||||
XM_006711498.3 | XP_006711561.1 | 14 | 3255 | |||
XM_011509926.2 | XP_011508228.1 | 14 | 3255 | |||
XM_011509927.2 | XP_011508229.1 | 13 | 3228 | |||
XM_011509928.2 | XP_011508230.1 | 14 | 3162 | |||
XM_011509929.1 | XP_011508231.1 | 14 | 1651 | |||
XM_011509930.2 | XP_011508232.1 | 14 | 3255 | |||
XM_011509931.1 | XP_011508233.1 | 14 | 1601 | |||
XM_011509932.2 | XP_011508234.1 | 14 | 1534 | |||
XM_011509933.1 | XP_011508235.1 | 15 | 1883 | |||
XM_011509934.1 | XP_011508236.1 | 14 | 1638 | |||
XM_011509935.1 | XP_011508237.1 | 14 | 1794 | |||
XM_011509936.1 | XP_011508238.1 | 13 | 1535 | |||
XM_017002206.1 | XP_016857695.1 | 13 | 1608 | |||
GenBank mRNA | AK074796.1 | - | 11 | 1032 | ||
AK223278.1 | - | 14 | 1610 | |||
AK298592.1 | - | 12 | 1272 | |||
AK298914.1 | - | 4 | 346 | |||
AK314337.1 | - | 13 | 1620 | |||
BC018786.1 | - | 5 | 499 | |||
BC113846.1 | - | 13 | 1598 | 40008346 | ||
BC113847.1 | - | 14 | 1614 | 40008343 | ||
L38969.1 | - | 14 | 1612 |