Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252608.1 | NP_001239537.1 | 3 | 403 | ||
NM_007112.4 | NP_009043.1 | 5 | 763 | |||
NR_045553.1 | 4 | 667 | ||||
XM_006711498.3 | XP_006711561.1 | 5 | 2322 | |||
XM_011509926.2 | XP_011508228.1 | 5 | 2322 | |||
XM_011509928.2 | XP_011508230.1 | 5 | 2229 | |||
XM_011509929.1 | XP_011508231.1 | 5 | 718 | |||
XM_011509930.2 | XP_011508232.1 | 5 | 2322 | |||
XM_011509931.1 | XP_011508233.1 | 5 | 668 | |||
XM_011509932.2 | XP_011508234.1 | 5 | 601 | |||
XM_011509933.1 | XP_011508235.1 | 6 | 950 | |||
XM_011509934.1 | XP_011508236.1 | 5 | 705 | |||
XM_011509935.1 | XP_011508237.1 | 5 | 861 | |||
XM_011509936.1 | XP_011508238.1 | 4 | 602 | |||
XM_017002206.1 | XP_016857695.1 | 4 | 602 | |||
GenBank mRNA | AK074796.1 | - | 2 | 99 | ||
AK223278.1 | - | 5 | 677 | |||
AK298592.1 | - | 3 | 339 | |||
AK298803.1 | - | 3 | 321 | |||
BC113846.1 | - | 4 | 585 | 40008346 | ||
BC113847.1 | - | 5 | 681 | 40008343 | ||
L38969.1 | - | 5 | 679 |