Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306212.1 | NP_001293141.1 | 12 | 1467 | ||
NM_001306213.1 | NP_001293142.1 | 12 | 1519 | |||
NM_001306214.1 | NP_001293143.1 | 11 | 1461 | |||
NM_003248.5 | NP_003239.2 | 11 | 1633 | |||
XM_017009798.1 | XP_016865287.1 | 11 | 1613 | |||
XM_017009799.1 | XP_016865288.1 | 11 | 1613 | |||
XM_017009800.1 | XP_016865289.1 | 11 | 1613 | |||
GenBank mRNA | AK302842.1 | - | 11 | 1461 | ||
AK313587.1 | - | 11 | 1449 | |||
BC050456.1 | - | 12 | 1717 | 6191808 | ||
Z19585.1 | - | 11 | 1421 |