Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000460.3 | NP_000451.1 | 6 | 952 | ||
NM_001177597.2 | NP_001171068.1 | 6 | 940 | |||
NM_001177598.2 | NP_001171069.1 | 6 | 935 | |||
NM_001289997.1 | NP_001276926.1 | 7 | 836 | |||
NM_001289998.1 | NP_001276927.1 | 7 | 1219 | |||
NM_001290003.1 | NP_001276932.1 | 7 | 1219 | |||
NM_001290022.1 | NP_001276951.1 | 7 | 1207 | |||
NM_001290026.1 | NP_001276955.1 | 7 | 1202 | |||
NM_001290027.1 | NP_001276956.1 | 8 | 1103 | |||
NM_001290028.1 | NP_001276957.1 | 6 | 916 | |||
XM_017007107.1 | XP_016862596.1 | 8 | 1091 | |||
GenBank mRNA | AB102890.1 | - | 4 | 656 | ||
AK311002.1 | - | 4 | 346 | |||
BC130322.1 | - | 6 | 872 | 40146353 | ||
BC136338.1 | - | 6 | 884 | 9020325 | ||
BC143982.1 | - | 6 | 872 | 9052500 | ||
D32047.1 | - | 5 | 774 | |||
L33410.1 | - | 6 | 888 | |||
L36052.1 | - | 4 | 673 | |||
U11025.1 | - | 5 | 708 | |||
U42977.1 | - | 2 | 127 | |||
U59493.1 | - | 4 | 673 | |||
U59494.1 | - | 4 | 673 | |||
U59495.1 | - | 5 | 556 |