Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022037.2 | NP_071320.2 | 6 | 617 | ||
NM_022173.2 | NP_071505.2 | 7 | 650 | |||
XM_005264527.1 | XP_005264584.1 | 6 | 627 | |||
XM_005264528.4 | XP_005264585.1 | 6 | 562 | |||
XM_005264531.1 | XP_005264588.1 | 8 | 697 | |||
XM_011533077.1 | XP_011531379.1 | 7 | 660 | |||
XM_011533078.1 | XP_011531380.1 | 6 | 4323 | |||
XM_011533081.2 | XP_011531383.1 | 7 | 1423 | |||
XM_011533082.2 | XP_011531384.1 | 5 | 4995 | |||
XM_017004789.1 | XP_016860278.1 | 7 | 479 | |||
XM_017004790.1 | XP_016860279.1 | 6 | 445 | |||
XM_017004791.1 | XP_016860280.1 | 5 | 529 | |||
XM_017004792.1 | XP_016860281.1 | 6 | 2834 | |||
XM_017004793.1 | XP_016860282.1 | 6 | 2659 | |||
XM_017004794.1 | XP_016860283.1 | 6 | 1391 | |||
XM_017004795.1 | XP_016860284.1 | 8 | 657 | |||
XM_017004796.1 | XP_016860285.1 | 6 | 4165 | |||
XM_017004797.1 | XP_016860286.1 | 7 | 1961 | |||
XM_017004798.1 | XP_016860287.1 | 7 | 692 | |||
XM_017004799.1 | XP_016860288.1 | 7 | 654 | |||
XM_017004800.1 | XP_016860289.1 | 6 | 621 | |||
GenBank mRNA | AB209211.1 | - | 7 | 661 | ||
BC015944.1 | - | 7 | 457 | 3909177 | ||
CN295707.1 | - | 6 | 533 | |||
CX867570.1 | - | 7 | 623 | |||
M77142.1 | - | 6 | 456 |