Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033925.1 | NP_001029097.1 | 7 | 1129 | ||
NM_001323964.1 | NP_001310893.1 | 8 | 1454 | |||
NM_001323965.1 | NP_001310894.1 | 8 | 1197 | |||
NM_001323967.1 | NP_001310896.1 | 8 | 1146 | |||
NM_001323968.1 | NP_001310897.1 | 8 | 1191 | |||
NM_001323969.1 | NP_001310898.1 | 8 | 1140 | |||
NM_001323970.1 | NP_001310899.1 | 8 | 1191 | |||
NM_003252.3 | NP_003243.1 | 7 | 1078 | |||
NR_136661.1 | 7 | 1078 | ||||
NR_136662.1 | 8 | 1197 | ||||
NR_136663.1 | 8 | 1140 | ||||
XM_017016606.1 | XP_016872095.1 | 4 | 1082 | |||
GenBank mRNA | AB209260.1 | - | 3 | 1888 | ||
AK290695.1 | - | 7 | 1031 | |||
AK290984.1 | - | 7 | 728 | |||
AK291239.1 | - | 7 | 711 | |||
AK295243.1 | - | 8 | 591 | |||
AL833106.1 | - | 7 | 1014 | |||
AY633610.1 | - | 7 | 522 | |||
BC030025.1 | - | 8 | 1459 | 4821759 | ||
D64015.1 | - | 7 | 554 | |||
M96954.1 | - | 7 | 559 |