Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000362.4 | NP_000353.1 | 3 | 1471 | ||
GenBank mRNA | AK298126.1 | - | 4 | 291 | ||
AK314871.1 | - | 3 | 580 | |||
BC014277.2 | - | 3 | 528 | 3689386 | ||
BT006848.1 | - | 3 | 285 | |||
CR456593.1 | - | 3 | 333 | |||
L15078.1 | - | 3 | 243 | |||
S78453.1 | - | 3 | 561 | |||
U02571.1 | - | 3 | 566 | |||
U14394.1 | - | 3 | 572 | |||
U67195.1 | - | 3 | 563 | |||
X76227.1 | - | 3 | 355 |