Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001064.3 | NP_001055.1 | 2 | 314 | ||
NM_001135055.2 | NP_001128527.1 | 2 | 314 | |||
NM_001258028.1 | NP_001244957.1 | 2 | 314 | |||
NR_047580.1 | 2 | 314 | ||||
XM_011534054.1 | XP_011532356.1 | 2 | 252 | |||
XM_011534055.1 | XP_011532357.1 | 2 | 238 | |||
GenBank mRNA | AK093667.1 | - | 2 | 222 | ||
AK223614.1 | - | 2 | 222 | |||
AK289454.1 | - | 2 | 190 | |||
AK293438.1 | - | 2 | 222 | |||
AK301231.1 | - | 2 | 222 | |||
AK303191.1 | - | 3 | 181 | |||
BC002433.2 | - | 2 | 213 | 3346980 | ||
BC009970.2 | - | 2 | 212 | 4310396 | ||
BX647248.1 | - | 2 | 200 | |||
CB139769.1 | - | 2 | 252 | |||
DB503248.1 | - | 2 | 314 | |||
EU794667.1 | - | 2 | 212 | |||
KU178361.1 | - | 2 | 142 | |||
KU178362.1 | - | 2 | 142 | |||
L12711.1 | - | 2 | 240 | |||
U55017.1 | - | 2 | 142 |