Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001064.3 | NP_001055.1 | 10, 11 | 1564 | ||
NM_001135055.2 | NP_001128527.1 | 10, 11 | 1564 | |||
NM_001258028.1 | NP_001244957.1 | 11, 12 | 1588 | |||
NR_047580.1 | 10, 11 | 1454 | ||||
XM_011534054.1 | XP_011532356.1 | 11, 12 | 1526 | |||
XM_011534055.1 | XP_011532357.1 | 11, 12 | 1613 | |||
GenBank mRNA | AK057092.1 | - | 4, 5 | 1393 | ||
AK092084.1 | - | 5, 6 | 2427 | |||
AK093667.1 | - | 9, 10 | 1223 | |||
AK223614.1 | - | 10, 11 | 1472 | |||
AK289454.1 | - | 10, 11 | 1440 | |||
AK293438.1 | - | 11, 12 | 1496 | |||
AK301231.1 | - | 10, 11 | 1362 | |||
AK303191.1 | - | 11, 12 | 1431 | |||
BC002433.2 | - | 11, 12 | 1353 | 3346980 | ||
BC008615.1 | - | 5, 6 | 708 | |||
BC009970.2 | - | 10, 11 | 1462 | 4310396 | ||
BC024026.2 | - | 8, 9 | 1133 | |||
BQ430230.1 | - | 1, 2 | 112 | |||
BX647248.1 | - | 10, 11 | 1450 | |||
EU794667.1 | - | 10, 11 | 1462 | |||
GQ472197.1 | - | 5, 6 | 708 | |||
KU178361.1 | - | 10, 11 | 1392 | |||
KU178363.1 | - | 3, 4 | 278 | |||
L12711.1 | - | 10, 11 | 1490 | |||
U55017.1 | - | 10, 11 | 1392 | |||
X67688.1 | - | 11, 12 | 1462 |