Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003265.2 | NP_003256.1 | 4 | 2116 | ||
XM_017008577.1 | XP_016864066.1 | 2 | 1806 | |||
GenBank mRNA | AB445631.1 | - | 3 | 2015 | ||
AK302143.1 | - | 2 | 1594 | |||
AK314208.1 | - | 4 | 2115 | |||
BC043204.2 | - | 1 | 2098 | |||
BC059372.1 | - | 4 | 2115 | |||
BC094737.1 | - | 4 | 2115 | 30531515 | ||
BC096333.1 | - | 4 | 2093 | 40008349 | ||
BC096334.3 | - | 4 | 2093 | 40008350 | ||
BC096335.1 | - | 4 | 2093 | 40008351 | ||
DQ445682.1 | - | 4 | 2086 | |||
DQ470075.1 | - | 1 | 370 | |||
DQ470076.1 | - | 1 | 370 | |||
DQ470077.1 | - | 1 | 370 | |||
DQ470078.1 | - | 1 | 370 | |||
DQ470079.1 | - | 1 | 370 | |||
U88879.1 | - | 4 | 2088 |