Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004616.2 | NP_004607.1 | 4 | 341 | ||
XM_006719583.3 | XP_006719646.1 | 4 | 295 | |||
XM_017019913.1 | XP_016875402.1 | 3 | 377 | |||
GenBank mRNA | AK223022.1 | - | 3 | 269 | ||
AK313112.1 | - | 3 | 324 | |||
BC005246.1 | - | 4 | 341 | 3934330 | ||
BC070168.1 | - | 8 | 715 | |||
M35252.1 | - | 3 | 299 |