Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004617.3 | NP_004608.1 | 3, 4 | 708 | ||
GenBank mRNA | AK055577.1 | - | 3, 4 | 1315 | ||
AK315464.1 | - | 3, 4 | 595 | |||
BC001386.2 | - | 3, 4 | 578 | 3051146 | ||
BT007430.1 | - | 3, 4 | 411 | |||
U31449.1 | - | 3, 4 | 576 |